This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03101848_m1
See other EGFL6 GE Assays ›
Gene Symbol:
egfl6
Gene Name:
EGF-like-domain, multiple 6
Gene Aliases:
cb3, maeg, sb:cb3, wu:fc66b04, zgc:92328
Chromosome Location:
Chr.9: 54629920 - 54679574 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03101848_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: egfl6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002457.1 NP_001002457.1 7 - 8 810 58
XM_017357664.2 XP_017213153.1 7 - 8 883 58
XM_021478639.1 XP_021334314.1 6 - 7 720 58
GenBank mRNA BC076013.1 - 7 - 8 810 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of Notch signaling pathway
positive regulation of cell-substrate adhesion
notochord development
positive regulation of sprouting angiogenesis
integrin binding
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header