This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03100790_g1
See other SH3GL1B GE Assays ›
Gene Symbol:
sh3gl1b
Gene Name:
SH3-domain GRB2-like 1b
Gene Aliases:
cb831, sh3gl1
Chromosome Location:
Chr.8: 197 - 37703 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03100790_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sh3gl1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201497.1 NP_958905.1 4 - 5 570 67
XM_005166793.4 XP_005166850.1 4 - 5 632 67
XM_009303889.3 XP_009302164.1 4 - 5 632 67
XM_021478204.1 XP_021333879.1 4 - 5 686 67
GenBank mRNA BC045942.1 - 4 - 5 570 67
BC164167.1 - 4 - 5 370 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0