This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03100754_m1
See other ETF1B GE Assays ›
Gene Symbol:
etf1b
Gene Name:
eukaryotic translation termination factor 1b
Gene Aliases:
eRF1, etf1, hm:zeh0057, wu:fa19e07, wu:fi31f01, zeh0057
Chromosome Location:
Chr.10: 297507 - 306559 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03100754_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: etf1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201460.2 NP_958868.1 4 - 5 589 137
XM_021479101.1 XP_021334776.1 4 - 5 137
GenBank mRNA AY398343.1 - 4 - 5 589 137
BC066583.1 - 4 - 5 137

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation release factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic translational termination
translational termination
brain development
translation release factor activity
translation release factor activity, codon specific
sequence-specific mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0