This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03100573_m1
See other ZNF143B GE Assays ›
Gene Symbol:
znf143b
Gene Name:
zinc finger protein 143b
Gene Aliases:
Zfp143, zgc:66276, znf143
Chromosome Location:
Chr.18: 16924871 - 16937130 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03100573_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: znf143b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200979.1 NP_957273.1 9 - 10 986 102
XM_005159072.4 XP_005159129.1 10 - 11 1034 102
XM_005159073.4 XP_005159130.1 10 - 11 1022 102
XM_009293545.3 XP_009291820.1 10 - 11 1540 102
GenBank mRNA AY331976.1 - 10 - 11 1030 102
BC055577.1 - 9 - 10 986 102
BC124735.1 - 10 - 11 1541 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
central nervous system development
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0