This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03100112_g1
See other EPS8A GE Assays ›
Gene Symbol:
eps8a
Gene Name:
epidermal growth factor receptor pathway substrate 8a
Gene Aliases:
eps8, zgc:56041
Chromosome Location:
Chr.4: 151505 - 178510 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03100112_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eps8a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200242.1 NP_956536.1 10 - 11 1104 70
XM_005168496.3 XP_005168553.1 9 - 10 970 70
XM_009299913.3 XP_009298188.1 10 - 11 1010 70
XM_009299914.3 XP_009298189.1 10 - 11 1010 70
XM_009299915.2 XP_009298190.1 10 - 11 1010 70
XM_009299916.3 XP_009298191.1 10 - 11 1010 70
XM_009299917.2 XP_009298192.1 10 - 11 1010 70
XM_009299918.3 XP_009298193.1 10 - 11 1010 70
XM_009299919.3 XP_009298194.1 10 - 11 1010 70
XM_009299920.3 XP_009298195.1 10 - 11 1010 70
XM_009299921.2 XP_009298196.1 10 - 11 1010 70
XM_009299922.3 XP_009298197.1 9 - 10 970 70
XM_009299923.3 XP_009298198.1 10 - 11 1098 70
XM_017354975.2 XP_017210464.1 10 - 11 1010 70
XM_017354976.2 XP_017210465.1 10 - 11 1010 70
XM_021474970.1 XP_021330645.1 10 - 11 1149 70
GenBank mRNA BC047814.1 - 10 - 11 1104 70
BC165198.1 - 9 - 10 937 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
regulation of Rho protein signal transduction
barbed-end actin filament capping
actin filament bundle assembly
positive regulation of ruffle assembly
actin binding
Rac guanyl-nucleotide exchange factor activity
signaling adaptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0