This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03099775_g1
See other CTPS1A GE Assays ›
Gene Symbol:
ctps1a
Gene Name:
CTP synthase 1a
Gene Aliases:
cb1040, ctps, ctps1, ctpsa, wu:fb49e03, wu:fe17b03
Chromosome Location:
Chr.19: 16016037 - 16027093 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03099775_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ctps1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199211.2 NP_954681.2 14 - 15 1597 102
GenBank mRNA BC050512.1 - 1312 102
BC058048.1 - 14 - 15 1530 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

pyrimidine nucleotide biosynthetic process
CTP biosynthetic process
glutamine metabolic process
pyrimidine nucleobase biosynthetic process
notochord development
'de novo' CTP biosynthetic process
nucleotide binding
CTP synthase activity
ATP binding
ligase activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0