This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03099587_m1
See other EPB41B GE Assays ›
Gene Symbol:
epb41b
Gene Name:
erythrocyte membrane protein band 4.1b
Gene Aliases:
P4.1R, epb41, wu:fb70c02
Chromosome Location:
Chr.16: 26449743 - 26488169 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03099587_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: epb41b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175084.2 NP_778259.1 26 - 27 4722 66
GenBank mRNA AY124488.1 - 26 - 27 4722 66
BC162616.1 - 26 - 27 4650 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

erythrocyte differentiation
cortical actin cytoskeleton organization
actomyosin structure organization
embryonic hemopoiesis
protein binding
calmodulin binding
cytoskeletal protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0