This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03099492_g1
See other MYO7AA GE Assays ›
Gene Symbol:
myo7aa
Gene Name:
myosin VIIAa
Gene Aliases:
myo7a
Chromosome Location:
Chr.18: 3367553 - 3464355 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03099492_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: myo7aa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152983.1 NP_694515.1 22 - 23 3066 95
XM_021467573.1 XP_021323248.1 23 - 24 3217 95
GenBank mRNA AJ404001.1 - 22 - 23 3066 95
AJ404002.1 - 21 - 22 2946 95
BC163570.1 - 21 - 22 2954 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
actin filament organization
sensory organ development
sensory perception of sound
vesicle transport along actin filament
hair cell differentiation
detection of mechanical stimulus involved in sensory perception
inner ear receptor stereocilium organization
protein localization to cilium
microfilament motor activity
nucleotide binding
motor activity
actin binding
ATP binding
actin-dependent ATPase activity
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0