This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03099331_g1
See other ELAVL1A GE Assays ›
Gene Symbol:
elavl1a
Gene Name:
ELAV like RNA binding protein 1a
Gene Aliases:
elavl1, elra, fa25f08, fb59a02, fc60g02, hua, wu:fa25f08, wu:fb59a02, wu:fc60g02, zHuA
Chromosome Location:
Chr.2: 37123338 - 37134217 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03099331_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: elavl1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131452.1 NP_571527.1 4 - 5 481 87
XM_005171267.4 XP_005171324.1 5 - 6 612 87
XM_005171268.4 XP_005171325.1 6 - 7 606 87
XM_009298634.2 XP_009296909.1 4 - 5 482 87
XM_017357655.1 XP_017213144.1 5 - 6 737 87
GenBank mRNA AF184244.1 - 4 - 5 481 87
BC044184.1 - 5 - 6 564 87
U17600.1 - 2 - 3 188 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

erythrocyte differentiation
positive regulation of angiogenesis
nucleic acid binding
RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0