This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03097342_m1
See other TIGARA GE Assays ›
Gene Symbol:
tigara
Gene Name:
TP53 induced glycolysis regulatory phosphatase a
Gene Aliases:
c12orf5a, zgc:136900
Chromosome Location:
Chr.25: 16962593 - 16969922 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03097342_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tigara
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039836.1 NP_001034925.1 5 - 6 541 65
XM_021470524.1 XP_021326199.1 4 - 5 264 65
XM_021470525.1 XP_021326200.1 3 - 4 299 65
GenBank mRNA BC114302.1 - 5 - 6 541 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

fructose 2,6-bisphosphate metabolic process
autophagy
apoptotic process
fructose 1,6-bisphosphate metabolic process
negative regulation of programmed cell death
regulation of pentose-phosphate shunt
positive regulation of DNA repair
negative regulation of glycolytic process
catalytic activity
bisphosphoglycerate 2-phosphatase activity
fructose-2,6-bisphosphate 2-phosphatase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0