This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096721_m1
See other DNMT3AA GE Assays ›
Gene Symbol:
dnmt3aa
Gene Name:
DNA (cytosine-5-)-methyltransferase 3 alpha a
Gene Aliases:
dnmt8
Chromosome Location:
Chr.20: 46967269 - 47099943 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096721_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dnmt3aa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001018134.1 NP_001018144.1 19 - 20 2334 68
XM_005160858.4 XP_005160915.1 22 - 23 3229 68
GenBank mRNA AB196919.1 - 19 - 20 2334 68
BC162629.1 - 18 - 19 2331 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
regulation of gene expression
methylation
methyltransferase activity
DNA-methyltransferase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header