This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096534_g1
See other GOLGA7BA GE Assays ›
Gene Symbol:
golga7ba
Gene Name:
golgin A7 family, member Ba
Gene Aliases:
golga7b, zgc:110549
Chromosome Location:
Chr.1: 54908900 - 54931913 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096534_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: golga7ba
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017566.2 NP_001017566.1 3 - 4 404 59
XM_005160170.4 XP_005160227.1 3 - 4 581 59
GenBank mRNA BC080249.1 - 3 - 4 584 59
BC092359.1 - 3 - 4 408 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to membrane
peptidyl-L-cysteine S-palmitoylation
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0