This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096393_gH
See other RAB6BB GE Assays ›
Gene Symbol:
rab6bb
Gene Name:
RAB6B, member RAS oncogene family b
Gene Aliases:
wu:fj49d11, zgc:112018
Chromosome Location:
Chr.2: 22795507 - 22801810 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096393_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rab6bb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013467.1 NP_001013485.1 3 - 4 325 87
GenBank mRNA BC091529.1 - 3 - 4 325 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
retrograde vesicle-mediated transport, Golgi to ER
intra-Golgi vesicle-mediated transport
retrograde transport, endosome to Golgi
GTPase activity
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0