Assay Details
Gene Symbol: gdf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012383.1 | NP_001012383.1 | 1 - 2 | 431 | 86 | |
GenBank mRNA | AY833104.1 | - | 1 - 2 | 431 | 86 | |
BC108013.1 | - | 1 - 2 | 429 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
