This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096329_m1
See other FGFRL1B GE Assays ›
Gene Symbol:
fgfrl1b
Gene Name:
fibroblast growth factor receptor like 1b
Gene Aliases:
im:7154242
Chromosome Location:
Chr.14: 41600339 - 41678813 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096329_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgfrl1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012263.2 NP_001012263.2 4 - 5 737 90
XM_005173306.4 XP_005173363.1 5 - 6 983 90
GenBank mRNA AJ781308.2 - 4 - 5 737 90
AJ781309.1 - 4 - 5 737 90
BC162498.1 - 4 - 5 737 90
CU458876.1 - 4 - 5 711 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cartilage development
fibroblast growth factor-activated receptor activity
protein binding
fibroblast growth factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0