This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096103_g1
See other NUDT3B GE Assays ›
Gene Symbol:
nudt3b
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 3b
Gene Aliases:
zgc:103443
Chromosome Location:
Chr.6: 54154790 - 54165246 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096103_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nudt3b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006034.2 NP_001006034.1 3 - 4 413 93
XM_017356473.2 XP_017211962.1 3 - 4 430 93
GenBank mRNA BC083382.1 - 3 - 4 414 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

diphosphoinositol polyphosphate metabolic process
diadenosine pentaphosphate catabolic process
diadenosine hexaphosphate catabolic process
adenosine 5'-(hexahydrogen pentaphosphate) catabolic process
endopolyphosphatase activity
diphosphoinositol-polyphosphate diphosphatase activity
hydrolase activity
bis(5'-adenosyl)-hexaphosphatase activity
bis(5'-adenosyl)-pentaphosphatase activity
m7G(5')pppN diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0