This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096073_m1
See other PIP5K1BA GE Assays ›
Gene Symbol:
pip5k1ba
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase, type I, beta a
Gene Aliases:
pip5k1b, zgc:101046
Chromosome Location:
Chr.10: 15881305 - 15919839 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096073_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pip5k1ba
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005775.1 NP_001005775.1 13 - 14 1695 68
XM_005155571.4 XP_005155628.1 14 - 15 68
XM_005155572.4 XP_005155629.1 13 - 14 68
XM_009305190.3 XP_009303465.1 14 - 15 68
XM_009305191.3 XP_009303466.1 14 - 15 68
GenBank mRNA BC083170.1 - 13 - 14 1695 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
phosphatidylinositol metabolic process
nucleotide binding
ATP binding
kinase activity
phosphatidylinositol phosphate kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0