This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03096065_gH
See other CTHL GE Assays ›
Gene Symbol:
cthl
Gene Name:
cystathionase (cystathionine gamma-lyase), like
Gene Aliases:
zgc:92377
Chromosome Location:
Chr.6: 37389683 - 37403444 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03096065_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cthl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001005400.1 NP_001005400.1 9 - 10 1041 68
GenBank mRNA BC080251.1 - 9 - 10 1041 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

cysteine biosynthetic process via cystathionine
transsulfuration
catalytic activity
cystathionine gamma-lyase activity
lyase activity
carbon-sulfur lyase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0