This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03095924_m1
See other GIPC1 GE Assays ›
Gene Symbol:
gipc1
Gene Name:
GIPC PDZ domain containing family, member 1
Gene Aliases:
GIPC, TIP-2, rgs19ip1
Chromosome Location:
Chr.3: 49125026 - 49137955 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03095924_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gipc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004018.1 NP_001004018.1 1 - 2 306 75
XM_005163776.4 XP_005163833.1 2 - 3 597 75
GenBank mRNA AY690668.1 - 1 - 2 306 75
AY989813.1 - 1 - 2 306 75
BC163671.1 - 1 - 2 350 75
BC163674.1 - 1 - 2 350 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
lymphangiogenesis
artery morphogenesis
venous blood vessel morphogenesis
MAP kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0