This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03094292_m1
See other STIMATE GE Assays ›
Gene Symbol:
stimate
Gene Name:
STIM activating enhance
Gene Aliases:
tmem110, zgc:63630
Chromosome Location:
Chr.11: 192939 - 206508 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03094292_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: stimate
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200467.1 NP_956761.1 4 - 5 668 84
GenBank mRNA BC055172.1 - 4 - 5 668 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

activation of store-operated calcium channel activity
calcium-mediated signaling using intracellular calcium source
calcium channel regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0