This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03094162_g1
See other CALCOCO1B GE Assays ›
Gene Symbol:
calcoco1b
Gene Name:
calcium binding and coiled-coil domain 1b
Gene Aliases:
tceb3, wu:fw66g10, zgc:56638
Chromosome Location:
Chr.11: 2111582 - 2131458 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03094162_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: calcoco1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200121.2 NP_956415.2 10 - 11 1386 85
XM_017358207.2 XP_017213696.1 10 - 11 1562 85
XM_021479558.1 XP_021335233.1 10 - 11 1564 85
GenBank mRNA BC051625.1 - 10 - 11 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

translation
translational elongation
positive regulation of transcription from RNA polymerase II promoter
transcription coactivator activity
translation elongation factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0