This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03093913_m1
See other MTHFD2L GE Assays ›
Gene Symbol:
mthfd2l
Gene Name:
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2 like
Gene Aliases:
wu:fi14c12, zgc:63479
Chromosome Location:
Chr.8: 32389649 - 32405069 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03093913_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: mthfd2l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199813.1 NP_956107.1 1 - 2 183 75
XM_005167146.4 XP_005167203.1 1 - 2 372 75
GenBank mRNA BC055121.1 - 1 - 2 183 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

one-carbon metabolic process
tetrahydrofolate interconversion
oxidation-reduction process
catalytic activity
methenyltetrahydrofolate cyclohydrolase activity
methylenetetrahydrofolate dehydrogenase (NAD+) activity
methylenetetrahydrofolate dehydrogenase (NADP+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0