This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03093600_m1
See other P2RX4A GE Assays ›
Gene Symbol:
p2rx4a
Gene Name:
purinergic receptor P2X, ligand-gated ion channel, 4a
Gene Aliases:
p2rx4, p2x4, wu:fi03f02
Chromosome Location:
Chr.21: 15723066 - 15732401 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03093600_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: p2rx4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153653.1 NP_705939.1 11 - 12 1173 115
XM_009295279.3 XP_009293554.1 10 - 11 951 116
GenBank mRNA AF317643.1 - 11 - 12 1173 115
BC042317.1 - 11 - 12 1333 116
BC066495.1 - 11 - 12 1162 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

ion transport
cation transport
calcium ion transport
response to ATP
cation transmembrane transport
nucleotide binding
purinergic nucleotide receptor activity
CTP binding
extracellular ATP-gated cation channel activity
ion channel activity
ATP binding
ligand-gated ion channel activity
purine nucleotide binding
ATP-gated ion channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0