This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03092557_g1
See other HIP1RB GE Assays ›
Gene Symbol:
hip1rb
Gene Name:
huntingtin interacting protein 1 related b
Gene Aliases:
hip1r, zgc:158784
Chromosome Location:
Chr.5: 66366865 - 66429357 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03092557_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hip1rb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083565.2 NP_001077034.2 22 - 23 2516 70
GenBank mRNA BC133158.1 - 22 - 23 2515 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
actin filament organization
positive regulation of receptor-mediated endocytosis
clathrin coat assembly
regulation of clathrin-mediated endocytosis
actin binding
phospholipid binding
clathrin binding
clathrin light chain binding
phosphatidylinositol binding
clathrin adaptor activity
phosphatidylinositol-3,4-bisphosphate binding
actin filament binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0