This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03092271_m1
See other CAMK1DB GE Assays ›
Gene Symbol:
camk1db
Gene Name:
calcium/calmodulin-dependent protein kinase 1Db
Gene Aliases:
camk1d, zgc:158713
Chromosome Location:
Chr.25: 7990207 - 8030118 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03092271_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: camk1db
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080658.2 NP_001074127.2 8 - 9 1134 66
NM_001135043.2 NP_001128515.1 8 - 9 1134 66
GenBank mRNA AB427148.1 - 8 - 9 839 66
AB427149.1 - 8 - 9 839 66
BC129402.1 - 8 - 9 1129 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
peptidyl-serine phosphorylation
chordate embryonic development
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0