Assay Details
Gene Symbol: eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077567.1 | NP_001071035.1 | 10 - 11 | 1524 | 63 | |
XM_021480133.1 | XP_021335808.1 | 10 - 11 | 1529 | 63 | ||
XM_021480134.1 | XP_021335809.1 | 10 - 11 | 1556 | 63 | ||
GenBank mRNA | BC124654.1 | - | 10 - 11 | 1524 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
