This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03091719_m1
See other PSTPIP1A GE Assays ›
Gene Symbol:
pstpip1a
Gene Name:
proline-serine-threonine phosphatase interacting protein 1a
Gene Aliases:
si:dkey-24l11.5
Chromosome Location:
Chr.18: 26930956 - 26963473 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03091719_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pstpip1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045216.1 NP_001038681.1 7 - 8 525 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament polymerization
cytoskeletal protein binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0