This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03091479_m1
See other ASAP2B GE Assays ›
Gene Symbol:
asap2b
Gene Name:
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2b
Gene Aliases:
fe36f09, si:dkey-81b15.3, wu:fe36f09, zgc:158353
Chromosome Location:
Chr.20: 29634611 - 29675457 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03091479_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: asap2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044933.2 NP_001038398.2 23 - 24 2701 102
XM_005160726.4 XP_005160783.1 23 - 24 2744 102
GenBank mRNA BC129209.1 - 23 - 24 2701 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of GTPase activity
GTPase activator activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0