This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03090602_g1
See other TENT2 GE Assays ›
Gene Symbol:
tent2
Gene Name:
terminal nucleotidyltransferase 2
Gene Aliases:
papd4, zgc:110560
Chromosome Location:
Chr.5: 51833137 - 51847896 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03090602_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tent2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001020600.1 NP_001018436.1 7 - 8 851 121
XM_021476157.1 XP_021331832.1 7 - 8 1018 121
GenBank mRNA BC095312.1 - 7 - 8 851 121

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
RNA stabilization
RNA polyadenylation
negative regulation of miRNA catabolic process
nucleotide binding
polynucleotide adenylyltransferase activity
ATP binding
transferase activity
nucleotidyltransferase activity
RNA polymerase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0