This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03090544_m1
See other CHRNA5 GE Assays ›
Gene Symbol:
chrna5
Gene Name:
cholinergic receptor, nicotinic, alpha 5
Gene Aliases:
zgc:110642
Chromosome Location:
Chr.18: 40486379 - 40508965 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03090544_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: chrna5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001017885.1 NP_001017885.1 8 - 9 1347 62
XM_021467652.1 XP_021323327.1 7 - 8 1843 62
GenBank mRNA BC092962.1 - 8 - 9 1347 62
BC164212.1 - 7 - 8 1274 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

ion transport
signal transduction
chemical synaptic transmission
synaptic transmission, cholinergic
ion transmembrane transport
response to nicotine
regulation of membrane potential
neurological system process
transmembrane signaling receptor activity
ion channel activity
extracellular ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation channel activity
neurotransmitter receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0