This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03090417_g1
See other SEMA3FA GE Assays ›
Gene Symbol:
sema3fa
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3Fa
Gene Aliases:
wu:fb92d12
Chromosome Location:
Chr.6: 42819009 - 42915774 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03090417_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: sema3fa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001014822.1 NP_001014822.1 2 - 3 294 118
XM_005166039.3 XP_005166096.1 3 - 4 839 118
XM_005166040.3 XP_005166097.1 3 - 4 839 118
GenBank mRNA AY766118.1 - 2 - 3 294 118
BC163764.1 - 2 - 3 350 118
BC163771.1 - 2 - 3 350 118
CU458859.1 - 2 - 3 309 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
axon guidance
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
semaphorin receptor binding
neuropilin binding
chemorepellent activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0