This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03089022_g1
See other LSM14AB GE Assays ›
Gene Symbol:
lsm14ab
Gene Name:
LSM14A mRNA processing body assembly factor b
Gene Aliases:
wu:fc55d12, zgc:55754, zgc:77202
Chromosome Location:
Chr.18: 302839 - 325512 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03089022_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: lsm14ab
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214772.2 NP_999937.2 5 - 6 851 132
XM_005158973.3 XP_005159030.1 6 - 7 950 132
XM_005158974.3 XP_005159031.1 6 - 7 950 132
GenBank mRNA AY577000.1 - 5 - 6 828 132
BC047185.1 - 5 - 6 848 132
BC065685.1 - 5 - 6 816 132
BC165583.1 - 5 - 6 710 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic mRNA processing body assembly
stress granule assembly
mRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0