This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03088952_mH
See other CYTH1A GE Assays ›
Gene Symbol:
cyth1a
Gene Name:
cytohesin 1a
Gene Aliases:
cyth1, fb38g09, pscd1, wu:fb38g09, zgc:152822
Chromosome Location:
Chr.3: 57462223 - 57578521 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03088952_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cyth1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_214682.2 NP_999847.1 5 - 6 549 76
XM_017353253.2 XP_017208742.1 5 - 6 636 76
XM_017353258.2 XP_017208747.1 5 - 6 467 76
GenBank mRNA AY574952.1 - 549 76
BC124139.1 - 5 - 6 542 76
BC164494.1 - 5 - 6 391 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of ARF protein signal transduction
epiboly involved in gastrulation with mouth forming second
ARF guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0