This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03087388_m1
See other SLC4A1A GE Assays ›
Gene Symbol:
slc4a1a
Gene Name:
solute carrier family 4 member 1a (Diego blood group)
Gene Aliases:
ae1, band3, si:dz180g5.1, slc4a1, zgc:111889, zgc:152771
Chromosome Location:
Chr.3: 20065861 - 20091960 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03087388_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: slc4a1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198338.1 NP_938152.1 12 - 13 1342 55
GenBank mRNA AF350072.1 - 12 - 13 1342 55
BC163932.1 - 12 - 13 1328 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
anion transport
bicarbonate transport
erythrocyte differentiation
embryonic hemopoiesis
ion homeostasis
regulation of intracellular pH
transmembrane transport
inorganic anion exchanger activity
protein binding
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
anion:anion antiporter activity
transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0