This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03087220_m1
See other RPS5 GE Assays ›
Gene Symbol:
rps5
Gene Name:
ribosomal protein S5
Gene Aliases:
chunp6933, fc74e05, fj57h03, wu:fc74e05, wu:fj57h03
Chromosome Location:
Chr.15: 39964943 - 39971704 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03087220_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rps5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173232.1 NP_775339.1 4 - 5 477 108
XM_021481330.1 XP_021337005.1 4 - 5 507 108
GenBank mRNA AF506223.1 - 4 - 5 477 108
BC059443.1 - 4 - 5 470 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

ribosomal small subunit assembly
translation
RNA binding
mRNA binding
structural constituent of ribosome
rRNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0