This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03087049_m1
See other EIF4EA GE Assays ›
Gene Symbol:
eif4ea
Gene Name:
eukaryotic translation initiation factor 4ea
Gene Aliases:
eif4e, eif4e-1, eif4e1a, zgc:86680
Chromosome Location:
Chr.14: 43577174 - 43586098 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03087049_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: eif4ea
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131733.1 NP_571808.1 6 - 7 537 89
GenBank mRNA AF257519.1 - 6 - 7 537 89
BC071364.1 - 6 - 7 550 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translation
RNA 7-methylguanosine cap binding
RNA binding
translation initiation factor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header