This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03086957_s1
See other FZD8B GE Assays ›
Gene Symbol:
fzd8b
Gene Name:
frizzled class receptor 8b
Gene Aliases:
ZG06, Zfz8b, fz6, fz8b, fzA, ik:tdsubc_1e1, xx:tdsubc_1e1, zfzA
Chromosome Location:
Chr.2: 76710 - 79215 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03086957_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fzd8b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131553.1 NP_571628.1 1 1849 90
GenBank mRNA AF039410.1 - 1 1842 90
AF060696.1 - 1 1849 90
AF117390.1 - 1 1663 90
BC067377.1 - 1 1852 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
multicellular organism development
dorsal/ventral pattern formation
Wnt signaling pathway
non-canonical Wnt signaling pathway
canonical Wnt signaling pathway
transmembrane signaling receptor activity
Wnt-protein binding
Wnt-activated receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0