This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03084210_m1
See other NEDD4A GE Assays ›
Gene Symbol:
nedd4a
Gene Name:
NEDD4 E3 ubiquitin protein ligase a
Gene Aliases:
si:ch211-79l17.5
Chromosome Location:
Chr.18: 507835 - 605949 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03084210_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: nedd4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034186.1 NP_001029358.1 2461 63
GenBank mRNA AY995154.1 - 2461 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Danio rerio

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
negative regulation of Notch signaling pathway
endothelial to hematopoietic transition
ubiquitin-protein transferase activity
transferase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0