This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03083515_g1
See other AOC2 GE Assays ›
Gene Symbol:
aoc2
Gene Name:
amine oxidase copper containing 2
Gene Aliases:
aoc3, fa94a08, fe14c05, wu:fa94a08, wu:fe14c05, zgc:113006
Chromosome Location:
Chr.3: 269492 - 272908 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03083515_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: aoc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013555.3 NP_001013573.3 3 - 4 2047 98
GenBank mRNA BC091681.1 - 3 - 4 810 98
BC164320.1 - 2 - 3 425 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

visual perception
amine metabolic process
oxidation-reduction process
copper ion binding
primary amine oxidase activity
oxidoreductase activity
metal ion binding
quinone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0