This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03081670_s1
See other GJA3 GE Assays ›
Gene Symbol:
gja3
Gene Name:
gap junction protein, alpha 3
Gene Aliases:
Cx44, cx46, cx48.5
Chromosome Location:
Chr.9: 273679 - 274983 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03081670_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gja3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207642.1 NP_997525.1 1 1249 65
XM_005167541.3 XP_005167598.1 3 1705 65
XM_005167542.3 XP_005167599.1 3 1420 65
XM_021478628.1 XP_021334303.1 3 1553 65
XM_021478629.1 XP_021334304.1 2 2076 65
GenBank mRNA AF465751.1 - 1 1249 65
BC163765.1 - 1 1305 65
BC163774.1 - 1 1305 65
GU363509.1 - 3 1418 65
GU363510.1 - 3 1411 65
GU363511.1 - 3 1539 65
GU363512.1 - 3 1691 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

cardiac conduction system development
cell communication
cell-cell signaling
visual perception
regulation of heart contraction
cell communication by electrical coupling
post-embryonic eye morphogenesis
cardiac muscle tissue morphogenesis
gap junction channel activity
gap junction hemi-channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0