This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03081231_m1
See other QKI2 GE Assays ›
Gene Symbol:
qki2
Gene Name:
QKI, KH domain containing, RNA binding 2
Gene Aliases:
qkib, qkic, qkr, zgc:65890, zgc:77340
Chromosome Location:
Chr.12: 34953072 - 35023266 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03081231_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: qki2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200842.2 NP_957136.2 4 - 5 1075 66
GenBank mRNA AB086654.1 - 4 - 5 1058 66
BC061709.1 - 4 - 5 991 66
BC065344.1 - 4 - 5 1075 66
BC155319.1 - 5 - 6 1057 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
regulation of translation
multicellular organism development
RNA splicing
skeletal myofibril assembly
cell differentiation
mRNA transport
nucleic acid binding
RNA binding
mRNA binding
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header