This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03081228_m1
See other QKI2 GE Assays ›
Gene Symbol:
qki2
Gene Name:
QKI, KH domain containing, RNA binding 2
Gene Aliases:
qkib, qkic, qkr, zgc:65890, zgc:77340
Chromosome Location:
Chr.12: 34953072 - 35023266 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03081228_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: qki2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_200842.2 NP_957136.2 1 - 2 669 81
XM_005156355.4 XP_005156412.1 1 - 2 585 81
XM_021480038.1 XP_021335713.1 1 - 2 779 81
XM_021480039.1 XP_021335714.1 1 - 2 781 81
GenBank mRNA AB086654.1 - 1 - 2 652 81
BC061709.1 - 1 - 2 585 81
BC065344.1 - 1 - 2 669 81
BC155319.1 - 2 - 3 651 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
regulation of translation
multicellular organism development
RNA splicing
skeletal myofibril assembly
cell differentiation
mRNA transport
nucleic acid binding
RNA binding
mRNA binding
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0