This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03080033_m1
See other FGF4 GE Assays ›
Gene Symbol:
fgf4
Gene Name:
fibroblast growth factor 4
Gene Aliases:
fc89b01, id:ibd5158, wu:fc89b01
Chromosome Location:
Chr.7: 54617076 - 54624873 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03080033_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fgf4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_131635.3 NP_571710.1 1 - 2 614 78
GenBank mRNA AF283555.1 - 1 - 2 294 78
BC128817.1 - 1 - 2 612 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
determination of left/right symmetry
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
organ morphogenesis
positive regulation of gene expression
cell differentiation
regulation of cell migration
cilium morphogenesis
Kupffer's vesicle development
fibroblast growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0