This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03079782_g1
See other GC2 GE Assays ›
Gene Symbol:
gc2
Gene Name:
guanylyl cyclase 2
Gene Aliases:
RETGC-2
Chromosome Location:
Chr.7: 51267545 - 51297609 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03079782_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109695.1 NP_001103165.1 14 - 15 2955 70
XM_017356806.2 XP_017212295.1 14 - 15 2955 70
XM_021477384.1 XP_021333059.1 15 - 16 3250 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
protein phosphorylation
signal transduction
receptor guanylyl cyclase signaling pathway
cyclic nucleotide biosynthetic process
intracellular signal transduction
nucleotide binding
peptide receptor activity
guanylate cyclase activity
protein kinase activity
ATP binding
lyase activity
phosphorus-oxygen lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0