This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03078005_m1
See other VEGFD GE Assays ›
Gene Symbol:
vegfd
Gene Name:
vascular endothelial growth factor D
Gene Aliases:
figf, vegf-d
Chromosome Location:
Chr.11: 29906384 - 29917432 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03078005_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: vegfd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040178.1 NP_001035268.1 4 - 5 616 78
XM_005169310.4 XP_005169367.1 4 - 5 1317 78
GenBank mRNA BC163164.1 - 4 - 5 669 78
BC163170.1 - 4 - 5 669 78
DQ402074.1 - 4 - 5 616 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
blood vessel development
response to hypoxia
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
lymph vessel development
lymphangiogenesis
sprouting angiogenesis
blood circulation
vascular endothelial growth factor signaling pathway
positive regulation of angiogenesis
vascular endothelial growth factor receptor signaling pathway
blood vessel morphogenesis
induction of positive chemotaxis
positive regulation of mast cell chemotaxis
vascular endothelial growth factor receptor binding
protein binding
growth factor activity
chemoattractant activity
vascular endothelial growth factor receptor 3 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0