This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03077961_g1
See other CDH10A GE Assays ›
Gene Symbol:
cdh10a
Gene Name:
cadherin 10, type 2a (T2-cadherin)
Gene Aliases:
cdh10
Chromosome Location:
Chr.2: 28889936 - 28934284 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03077961_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cdh10a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039633.1 NP_001034722.1 6 - 7 1392 97
GenBank mRNA DQ303467.1 - 6 - 7 1392 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cell-cell junction assembly
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
adherens junction organization
cell-cell adhesion
cell-cell adhesion via plasma-membrane adhesion molecules
calcium ion binding
cadherin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0