This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03077784_m1
See other CABP2A GE Assays ›
Gene Symbol:
cabp2a
Gene Name:
calcium binding protein 2a
Gene Aliases:
zgc:112300
Chromosome Location:
Chr.1: 44107820 - 44141442 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03077784_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cabp2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001030268.2 NP_001025439.2 5 - 6 548 94
XM_009293387.3 XP_009291662.1 5 - 6 655 94
GenBank mRNA BC096884.1 - 5 - 6 548 94

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons

Gene Ontology Categories:

Process(es)

calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0