This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03077098_m1
See other ARHGEF7B GE Assays ›
Gene Symbol:
arhgef7b
Gene Name:
Rho guanine nucleotide exchange factor (GEF) 7b
Gene Aliases:
arhgef7, bbh, bpix, im:7142367, wu:fc35h08, zgc:101775
Chromosome Location:
Chr.1: 45958725 - 46020725 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03077098_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: arhgef7b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008624.1 NP_001008624.1 16 - 17 2057 79
XM_017354714.2 XP_017210203.1 16 - 17 2236 79
XM_017354716.2 XP_017210205.1 16 - 17 1869 79
XM_017354718.2 XP_017210207.1 16 - 17 1832 79
XM_021474150.1 XP_021329825.1 16 - 17 1942 79
GenBank mRNA BC086736.1 - 16 - 17 2057 79
BC165668.1 - 16 - 17 1976 79
DQ656108.1 - 16 - 17 1843 79
DQ656109.1 - 16 - 17 2011 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
blood vessel development
endothelial cell proliferation
intracellular signal transduction
epiboly involved in gastrulation with mouth forming second
protein localization to adherens junction
guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0