This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03075936_m1
See other SYT11A GE Assays ›
Gene Symbol:
syt11a
Gene Name:
synaptotagmin XIa
Gene Aliases:
fc52d10, fi68d01, syt11, wu:fc52d10, wu:fi68d01, zgc:55556
Chromosome Location:
Chr.19: 23982416 - 24032417 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03075936_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: syt11a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_213460.2 NP_998625.2 2 - 3 1111 70
GenBank mRNA BC044424.1 - 2 - 3 1051 70
BC097077.1 - 2 - 3 1062 70
BC165339.1 - 2 - 3 960 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

vesicle fusion
regulation of dopamine secretion
vesicle-mediated transport
calcium ion regulated exocytosis
regulation of calcium ion-dependent exocytosis
cellular response to calcium ion
SNARE binding
phosphatidylserine binding
calcium ion binding
calcium-dependent phospholipid binding
clathrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0