This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Dr03075612_m1
See other CNOT6A GE Assays ›
Gene Symbol:
cnot6a
Gene Name:
CCR4-NOT transcription complex, subunit 6a
Gene Aliases:
cnot6, wu:fa03c11, wu:fc17f01, zgc:65822
Chromosome Location:
Chr.21: 32755467 - 32781612 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID Dr03075612_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cnot6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212660.2 NP_997825.1 4 - 5 491 114
XM_009295585.3 XP_009293860.1 4 - 5 476 114
GenBank mRNA BC050504.1 - 4 - 5 415 114
BC058309.1 - 4 - 5 414 114
CT690778.2 - 4 - 5 445 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA poly(A) tail shortening
3'-5'-exoribonuclease activity
poly(A)-specific ribonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0